Learn More
eMolecules​ p-Anisidine | Oakwood Chemical | 104-94-9 | MFCD00007864 | 123.155 | C7H9NO | 99.000 | COc1ccc(N)cc1 | 100g | 537691322
Supplier:  eMolecules​ 537691322
Encompass_Preferred
p-Anisidine | Oakwood Chemical | 104-94-9 | MFCD00007864 | 123.155 | C7H9NO | 99.000 | COc1ccc(N)cc1 | 100g | 537691322
Catalog No. NC2259050