Learn More
Medchemexpress LLC (-)-p-Bromotetramisole oxalate | 62284-79-1 | 99.9% | 373.22 | 50 MG
Supplier: Medchemexpress LLC HY1969550MG
(-)-p-Bromotetramisole oxalate is an alkaline phosphatase inhibitor applicable to research related to erythroleukemia.
- Inhibits terminal cell division-associated MEL DS-19 cell differentiation induced by DMSO or HMBA.
- Reduces B+ cell levels.
- Potently inhibits purified human liver, bone, spleen, and kidney alkaline phosphatase isoenzymes.
- Minimally inhibits purified human intestinal and placental alkaline phosphatase isoenzymes.