Learn More
Medchemexpress LLC P-CAB agent 2 | 1978371-23-1 | C22H25FN2O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY153219A5MG
Encompass_Preferred
P-CAB agent 2 is a potent and orally active potassium-competitive acid blocker and a gastric acid secretion inhibitor. It inhibits H+/K+-ATPase activity and shows no acute toxicity.
  • Inhibits H+/K+-ATPase activity with an IC50 value of <100 nM.
  • Inhibits the hERG potassium channel with an IC50 value of 18.69 μM.
  • Shows no acute toxicity.
  • Inhibits histamine-induced gastric acid secretion.
Catalog No. 50-003-72573