Learn More
Cayman Chemical p-chloro PhenylalnIn 5g
SDP
Supplier:  Cayman Chemical 261685 G
Encompass_Preferred
An inhibitor of tryptophan hydroxylase (Ki = 300 µM); a competitive inhibitor in vitro and an irreversible inhibitor in vivo; depletes serotonin to undetectable levels in rats after six days at 300 mg/kg; has commonly been used to deplete brain serotonin levels in animal studies of the serotonergic system
Catalog No. 50-293-3929