Learn More
Bioworld p-Chlorophenoxyacetic Acid (CPA), 100 g
Supplier:  Bioworld 306310072
Encompass_Preferred
p-Chlorophenoxyacetic acid (CPA)Synonym: 4-CPA Influence cell enlargement, root initiation and adventitious bud formation Appearance: off-white to tan crystals Melting Point: 157-159 °CSoluble in etanol and KOH Harmful if swallowed
Catalog No. 50-255-041