Learn More
Medchemexpress LLC p-Coumaryl alcohol | 3690-05-9 | 5 MG

Supplier: Medchemexpress LLC HYN31025MG
p-Coumaryl alcohol is an antioxidant isolated from Alpinia officinarum Hance, demonstrating inhibitory effects during the autoxidation of methyl linoleate. This white to off-white solid has a molecular weight of 150.17 and a chemical formula of C9H10O2. It is for research use only and not sold to patients.
- Antioxidant properties
- Isolated from Alpinia officinarum Hance
- Inhibitory effect on methyl linoleate autoxidation
- Molecular weight: 150.17
- Formula: C9H10O2
- Appears as a white to off-white solid
- Purity: 98.43%
- Storage: 4°C, under nitrogen; in solvent, -80°C for 6 months or -20°C for 1 month, under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.