Learn More
Medchemexpress LLC p-Coumaryl alcohol | 3690-05-9 | MFCD00018809 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN31021MG
Encompass_Preferred
p-Coumaryl alcohol is an antioxidant that can be isolated from Alpinia officinarum Hance. It exhibits an inhibitory effect during the autoxidation of methyl linoleate. This compound is a phenylpropanoid, acting as a crucial monolignol precursor in the biosynthesis of lignin, a complex organic polymer found in plant cell walls.
  • Antioxidant properties.
  • Isolated from Alpinia officinarum Hance.
  • Inhibits autoxidation of methyl linoleate.
  • Crucial monolignol precursor.
  • Involved in lignin biosynthesis.
Catalog No. 50-004-21900