Learn More
Medchemexpress LLC p-coumaryl alcohol (p-hydroxycinnamyl alcohol) | 3690-05-9 | MFCD02169461 | 98.4% | 150.17 g/mol | C9H10O2 | 10 MG

Supplier: Medchemexpress LLC HYN310210MG
p-Coumaryl alcohol is a phenolic antioxidant isolated from Alpinia officinarum Hance that inhibits the autoxidation of methyl linoleate. It is supplied as a research-grade chemical suitable for laboratory studies and analytical investigations.
- Antioxidant that inhibits autoxidation of methyl linoleate.
- High purity: 98.43%.
- Molecular formula C9H10O2; molecular weight 150.17 g/mol.
- White to off-white solid appearance.
- Storage: store under nitrogen; in solvent, -80°C for up to 6 months or -20°C for up to 1 month.
- Available in small laboratory quantities (e.g., 1 mg-100 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.