Learn More
Medchemexpress LLC p-cresol sulfate-d4 potassium | 91978-69-7 | 230.32 | C7H3D4KO4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW6541215MG
Encompass_Preferred
p-Cresol sulfate-d4 potassium is a deuterium-labeled potassium salt of p-cresyl sulfate used as an analytical reference standard. It serves as an internal standard or tracer in mass spectrometry and metabolic studies where isotopic labeling improves quantitation and identification. The product is supplied as a small solid quantity suitable for laboratory analysis.
  • Deuterated internal standard for mass spectrometry.
  • Improves quantitation and peak identification in LC-MS/MS workflows.
  • Supplied as a stable potassium salt for easy handling.
  • Provided in small, laboratory-scale quantities for analytical use.
  • Certificate of analysis available for purity and documentation.
Catalog No. 50-002-84719