Learn More
Medchemexpress LLC P-cresol sulfate-d7 potassium | 2469278-96-2 | MFCD06658145 | 99.6% | 233.33 | C7D7KO4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY111431AS5MG
Encompass_Preferred
p-Cresol sulfate-d7 potassium is the deuterium-labeled potassium salt of p-cresyl sulfate supplied as an isotopically labeled reference standard for analytical research. It is intended for use as a tracer and as an internal standard for quantitative NMR, GC-MS, or LC-MS, and is provided as a solid with high purity.
  • Deuterium-labeled potassium salt for isotopic standardization.
  • Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity suitable for quantitative analysis.
  • Solid form, recommended storage sealed at 4°C; in solvent store at -80°C for long-term stability.
  • Documentation available: COA, SDS, and data sheet for quality and handling information.
Catalog No. 50-002-63149