Learn More
Santa Cruz Biotechnology P-ERK
Supplier:  Santa Cruz Biotechnology SC7383
Encompass
p-ERK (E-4), mouse monoclonal IgG2a; 200ug/mL, Western blotting, immunoprecipitation and immunohistochemistry (including paraffin-embedded sections), agarose conjugate for immunoprecipitation studies
Catalog No. NC9553305