Learn More
Medchemexpress LLC HY-23460 50mg Medchemexpress, p-Ethynylphenylalanine CAS:278605-15-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY2346050MG
Encompass_Preferred
Medchemexpress, HY-23460 50mg p-Ethynylphenylalanine CAS:278605-15-5 p-Ethynylphenylalanine (4-Ethynyl-L-phenylalanine) is a potent, selective, reversible and competitive inhibitor of tryptophan hydroxylase (TPH), with a Ki of 32.6 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2221