Learn More
Medchemexpress LLC P-gp inhibitor 1 | 2050747-49-2 | 98.8% | 517.62 g/mol | C32H31N5O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10179150MG
Encompass_Preferred
P-gp inhibitor 1 is a small-molecule research compound that reverses P-glycoprotein-mediated multidrug resistance in vitro. It shows high potency and low cytotoxicity in cellular assays, making it useful for studies of transporter function and chemosensitization.
  • Reverses P-glycoprotein-mediated multidrug resistance in cell-based assays.
  • Demonstrates high potency (reported EC50 ≈ 57.9 nM).
  • Exhibits low cytotoxicity in reported studies.
  • Suitable for in vitro studies of transporter biology and chemosensitization.
  • Stable under recommended storage; protect from light.
Catalog No. 50-002-47722