Learn More
Medchemexpress LLC p-Hydroxymandelic acid | 1198-84-1 | 98.9% | 168.15 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY113027
Encompass_Preferred
p-Hydroxymandelic acid is a valuable aromatic fine chemical used widely for the production of pharmaceuticals and food additives. It is also utilized for the preparation of 4-hydroxyphenylacetic acid, which is a synthetic precursor for the selective β1-receptor antagonist drug atenolol. This compound can conjugate cytotoxic drugs and enzyme substrates, suggesting its application in targeted drug delivery systems.
  • Valuable aromatic fine chemical.
  • Widely used in pharmaceuticals and food additives.
  • Precursor for 4-hydroxyphenylacetic acid, which is used to synthesize atenolol.
  • Can be used in targeted drug delivery systems by conjugating cytotoxic drugs and enzyme substrates.
  • For research use only.
Catalog No. 50-004-39034