Learn More
Medchemexpress LLC p-Hydroxyphenethyl trans-ferulate | 84873-15-4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN3078
Encompass_Preferred
p-Hydroxyphenethyl trans-ferulate exhibits a range of biological activities including anti-hyperglycemic (yeast α-glucosidase, IC50=19.24 ± 1.73 μM), antioxidant, and anti-inflammatory properties. It also demonstrates inhibiting anticancer and serotonergic activity.
  • Anti-hyperglycemic activity
  • Antioxidant properties
  • Anti-inflammatory effects
  • Inhibits anticancer activity
  • Demonstrates serotonergic activity
Catalog No. 50-004-20531