Learn More
TARGETMOL CHEMICALS INC p-MPPF dihydrochloride 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T697501MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. p-MPPF dihydrochloride is a selective 5-HT antagonist that dose-dependently antagonizes hypothermia induced by 8-hydroxy-2-(di-n-propylamino)tetralin (8-OH-DPAT) and can be used to study neurological diseases. Purity 99.78%
Catalog No. 50-287-9970