Learn More
Medchemexpress LLC P-NH2-Bn-DOTA | 181065-46-3 | MFCD32661399 | 98.2% | C23H35N5O8 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY48876
Encompass_Preferred
P-NH2-Bn-DOTA is a bifunctional chelator (BFC) and a macrocyclic DOTA derivative. It is primarily used for tumor pre-targeting and can be utilized for the conjugation of peptides and radionuclides.
  • Bifunctional chelator (BFC)
  • Macrocyclic DOTA derivative
  • Supports tumor pre-targeting
  • Enables conjugation of peptides
  • Enables conjugation of radionuclides
  • Can be used in the preparation of gadolinium complexes
  • Useful as MRI blood contrast agents
Catalog No. 50-003-57962