Learn More
Medchemexpress LLC p-nitrobenzyl mesylate | 39628-94-9 | MFCD00014798 | ≥99.0% | 231.23 g/mol | C8H9NO5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1183805MG
Encompass_Preferred
p-Nitrobenzyl mesylate is a small-molecule alkylating reagent used to selectively modify thiols and phosphorothioates in biochemical assays, including kinase substrate identification. It is supplied as a low-mass solid or as a 10 mM solution in DMSO for assay applications.
  • Small-molecule alkylating reagent for thiol and phosphorothioate modification
  • Suitable for direct kinase substrate identification and biochemical assays
  • Available as a solid and as a 10 mM solution in DMSO for experimental flexibility
  • High purity compatible with sensitive analytical workflows
  • Well characterized molecular weight and formula for stoichiometric calculations
Catalog No. 50-002-26172