Learn More
Medchemexpress LLC p-nitrobenzyl mesylate | 39628-94-9 | MFCD25963099 | 98.8% | 231.23 | C8H9NO5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY118380100MG
Encompass_Preferred
p-Nitrobenzyl mesylate is a biochemical alkylating reagent used to modify phosphorothioates, producing phosphorothioate epitopes that can be detected by antibodies.
  • Reagent for alkylation of phosphorothioates for antibody-based detection.
  • Available as a solid (5-100 mg) and as a 10 mM solution in DMSO for convenient use.
  • High purity (≈98.8%), suitable for biochemical assays.
  • Analytical support files available, including COA, SDS, H NMR, and LCMS.
  • Used in kinase substrate identification and related biochemical workflows.
Catalog No. 50-002-28529