Learn More
Medchemexpress LLC P-nitrobenzyl mesylate | 39628-94-9 | MFCD25963099 | 98.8% | 231.23 g/mol | C8H9NO5S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11838025MG
Encompass_Preferred
p-Nitrobenzyl mesylate is a thiol-specific alkylating reagent used to alkylate thiophosphates and thiols in biochemical assays; it is commonly used for direct kinase substrate identification and is supplied as a solid or as DMSO solutions at research-grade purity.
  • Thiol-specific alkylating reagent for biochemical assays.
  • Used for direct kinase substrate identification.
  • High purity suitable for analytical and preparative work.
  • Molecular weight 231.23 g/mol; formula C8H9NO5S.
  • Available in small-scale quantities and as 10 mM DMSO solutions.
  • Product data sheet and safety data sheet available.
Catalog No. 50-002-27888