Learn More
Selleck Chemical LLC p-Nitrobenzyl mesylate-5MG
SDP
Supplier:  Selleck Chemical LLC E12485MG
Encompass_Preferred
p-Nitrobenzyl mesylate is an alkylating reagent for direct kinase substrate identification
Catalog No. 50-238-9689