Learn More
Crescent Chemical Co Inc P-NITROPHENOL SOLN 1ML
SDP
Supplier:  Crescent Chemical Co Inc CT3301M
Encompass_Preferred
p-Nitrophenol Single Component Calibration Standard in Methanol at 100 ug/mL 1mL
Catalog No. 50-689-41