Learn More
Alkali Scientific p Nitrophenyl β D mannopyranoside, 100 Mg
SDP
Supplier:  Alkali Scientific BDM001CP
Encompass
Nitrophenyl β-D-mannopyranoside is a reagent supplied in a 100mg quantity, widely used in biochemical research and enzyme activity assays. This compound serves as a substrate for β-mannosidase, an enzyme involved in the hydrolysis of mannopyranoside bonds. It is utilized in studying the enzymatic breakdown of mannan and related polysaccharides, which is important in plant biology, microbiology, and enzymology. The reagent is also used to analyze enzyme kinetics and to explore the role of β-mannosidase in carbohydrate metabolism. It is a valuable tool in research on glycosidic bond hydrolysis and the functional properties of glycosidases.
Catalog No. 50-006-60283