Learn More
Alkali Scientific p Nitrophenyl α D mannopyranoside, 500 Mg
SDP
Supplier:  Alkali Scientific LAD001CP
Encompass
p-Nitrophenyl α-D Mannopyranoside, 500mg, is a biochemical reagent used for studying α-mannosidase activity. This compound serves as a substrate for α-mannosidase, where the enzyme cleaves the bond between the p-nitrophenyl group and the α-D-mannopyranoside molecule, releasing a yellow product. This colorimetric reaction is commonly used in enzyme assays to study enzyme kinetics, carbohydrate metabolism, and for genetic research. The 500mg size is suitable for multiple experiments or larger-scale studies. It is commonly used in laboratory assays to monitor enzyme activity, particularly in microbiological studies and enzyme research, providing a reliable and simple method for enzyme analysis.
Catalog No. 50-006-59735