Learn More
Alkali Scientific p Nitrophenyl α L Arabinofuranoside, 1 G
Supplier: Alkali Scientific ALA004CP
Nitrophenyl α-L-arabinofuranoside is a biochemical reagent available in a 1g quantity for use in enzymatic assays and research. This substrate is commonly employed to measure the activity of α-arabinofuranosidase, an enzyme responsible for hydrolyzing arabinose-based oligosaccharides and polysaccharides. It is particularly useful in studies on carbohydrate metabolism, plant biochemistry, and the degradation of hemicellulose in plant cell walls. By using this reagent, researchers can investigate the enzymatic cleavage of arabinofuranoside bonds, which is vital in understanding complex carbohydrate breakdown processes in both plants and microorganisms.