Learn More
Alkali Scientific p Nitrophenyl a L fucopyranoside, 100 Mg
Supplier: Alkali Scientific PHE001CP
p-Nitrophenyl α-L-fucopyranoside is a chemical reagent commonly used in carbohydrate analysis. This 100 mg quantity is provided in powder form and is widely used in enzymatic assays, especially for studying enzymes involved in glycosidic bond cleavage. The product serves as a substrate in assays for enzymes like α-L-fucosidase. The compound is highly stable under standard laboratory conditions and is used in biochemical research to investigate fucose metabolism and related glycosidase activities. This reagent is essential for assays that require a reliable source of α-L-fucose for enzymatic testing.