Learn More
Cayman Chemical p-NItrophenyl Sulfatepotas 1g
SDP
Supplier:  Cayman Chemical 356911 G
Encompass_Preferred
A colorimetric arylsulfatase substrate; Upon enzymatic cleavage by arylsulfatase, p-nitrophenyl is released, which can be quantified by colorimetric detection at 400 nm as a measure of arylsulfatase activity
Catalog No. 50-291-1136