Learn More
Abcam p-Nitropifithrin-alpha, p53 inhibitor, 5MG
Supplier:  Abcam AB1458735MG
Encompass_Preferred
MW 398.3 Da, Purity >98%. Cell-permeable p53 inhibitor. Pifithrin-α (ab120478) analog and prodrug. Shows more potent antiapoptotic effects than pifithrin-α. Shows antiobesogenic and hepatoprotective effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5051