Learn More
Medchemexpress LLC P-SCN-Bn-deferoxamine | 1222468-90-7 | 98.0% | 752.94 g/mol | C33H52N8O8S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1347975MG
Encompass_Preferred
p-SCN-Bn-deferoxamine is a bifunctional chelator designed for bioconjugation and radiolabeling workflows. It couples to primary amines on proteins via a para-isothiocyanatobenzyl (p-SCN-Bn) reactive group and provides a deferoxamine chelator for binding radiometals, enabling preparation of antibody-based tracers and surface-functionalized biomaterials.
  • Reacts with primary amines via isothiocyanate chemistry.
  • Chelates radiometals for stable radiolabeling.
  • Suitable for antibody conjugation and PET tracer development.
  • Supplied as a white to off-white solid with high purity.
  • Stable when stored protected from light at recommended temperatures.
Catalog No. 50-002-23073