Learn More
Medchemexpress LLC p-SCN-Bn-DOTA | 5MG
SDP
Supplier:  Medchemexpress LLC HY1143775MG
Encompass_Preferred
p-SCN-Bn-DOTA is a bifunctional chelating agent p-SCN-Bn-DOTA can simultaneously chelate radionuclides and link monoclonal antibody for radioimmunoresearch of tumor[1][2]
Catalog No. 50-002-27481