Learn More
Medchemexpress LLC p-SCN-Bn-TCMC | 282097-63-6 | >98% | 547.67 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY201263100MG
Encompass_Preferred
p-SCN-Bn-TCMC is a bifunctional chelator. Its cyclen backbone binds Hg2+, while its isothiocyanate group allows for conjugation to biomolecules, making it suitable for various research applications. It can be used in the research of prostate cancer.
  • Bifunctional chelator
  • Cyclen backbone binds Hg2+
  • Isothiocyanate group allows for conjugation to biomolecules
  • Used in the research of prostate cancer
Catalog No. 50-004-04195