Learn More
Medchemexpress LLC P-SCN-Bn-TCMC tetrahydrochloride | 2780352-89-6 | >95.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY20126310MG
Encompass_Preferred
p-SCN-Bn-TCMC tetrahydrochloride is a bifunctional chelator that combines a TCMC macrocyclic chelating core with a para-isothiocyanatobenzyl reactive linker. It is intended for covalent attachment to biomolecules and subsequent radiometal complexation, enabling preparation of radiolabeled conjugates for imaging and therapeutic studies. The tetrahydrochloride salt improves solubility and handling.
Catalog No. 50-002-96880