Learn More
Strem, An Ascensus Company CAS# 1375325-71-5. 250mg. Chloro(tri-t-butylphosphine)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98%. MFCD21608496
SDP
Supplier:  Strem, An Ascensus Company 460028250MG
Encompass
CAS# 1375325-71-5. 250mg. Chloro(tri-t-butylphosphine)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98%. MFCD21608496. Molecular Weight: 512.40. Molecular Formula: C24H37ClNPPd. Color/form: yellow pwdr. Strem# 46-0028. http://www.strem.com/catalog/v/46-0028/
Catalog No. 50-204-5342