Learn More
Medchemexpress LLC P-Tolualdehyde-d7 | 1219805-23-8 | 98.2% | 127.19 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW012860S10MG
Encompass_Preferred
p-Tolualdehyde-d7 is the deuterium-labeled version of p-Tolualdehyde. p-Tolualdehyde is an active small molecule that can be used as a drug intermediate. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54879