Learn More
Medchemexpress LLC p-Tolualdehyde-d7 | 1219805-23-8 | 98.2% | 127.19 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW012860S100MG
Encompass_Preferred
p-Tolualdehyde-d7 is the deuterium-labeled p-Tolualdehyde. It is an active small molecule that can be used as a drug intermediate. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes like deuterium have been incorporated into drug molecules for quantitation during drug development and have the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-54942