Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC p-Toluenesulfonamide | 70-55-3 | 99.9% | C7H9NO2S | 100 G
SDP

Supplier:  Medchemexpress LLC HY79602100G

Encompass_Preferred

p-Toluenesulfonamide is an intermediate and plasticizer. It is also the main degradation product of the disinfectant Chloramine-T in water. This compound facilitates the localization of fluorescent probes to the endoplasmic reticulum.

  • Appears as a white to off-white solid
  • Molecular weight: 171.22
  • Purity: 99.98%
  • Soluble in DMSO (175 mg/mL)

Catalog No. 50-004-32722


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.