Learn More
New England Biolabs, Inc. NEB® Stable Competent E. coli (High Efficiency) – 20 x 0.05 ml/tube
SDP
Supplier:  New England Biolabs, Inc. C3040H
Encompass
Chemically competent E. coli cells suitable for high efficiency transformation and isolation of plasmid clones containing repeat elements.
  • Ideal for isolation of plasmid clones containing repeat elements and unstable inserts
  • Useful for isolating and propagating retroviral/lentiviral clones
  • Carries the endA mutation (in contrast to Stbl3) so that isolated plasmids are free of EndoI
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
Catalog No. 50-591-190
 Restricted Item