Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma hydrochloride solution, 1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T26941L
Encompass
pH 8.0, BioPerformance Certified, 1 M, suitable for cell culture, Synonym: Tris hydrochloride solution. Trizma hydrochloride solution has been used: as a component of extraction buffer for Plasmodiophora brassicae DNA isolation, as a component of resuspension buffer for human genomic DNA fragmented sample, and for adjusting pH of plasma samples and as a component of reconstitution and neutralization buffer.
Catalog No. NC1646206