Learn More
Medchemexpress LLC P11149, a galanthamine derivative | 164724-79-2 | 99.2% | 472.02 g·mol⁻1 | C27H34ClNO4 | 5 MG

Supplier: Medchemexpress LLC HY1053275MG
P11149 is a galanthamine-derived acetylcholinesterase (AChE) inhibitor used in preclinical pharmacology research, including Alzheimer's disease models. It is a competitive, orally active compound with weak blood-brain barrier penetration and reported IC50 ≈ 1.3 μM (rat BChE/AChE).
- Chemical formula: C27H34ClNO4.
- Molecular weight: 472.02 g·mol⁻1.
- Purity: 99.2%.
- Physical form: solid, white to off-white.
- Solubility: DMSO 100 mg/mL (ultrasonic recommended; hygroscopic DMSO affects solubility).
- Recommended storage: solid at 4 °C; in solvent -80 °C (6 months), -20 °C (1 month).
- Typical use: pharmacology studies of AChE inhibition and Alzheimer's disease models.
- CAS number: 164724-79-2.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.