Learn More
Medchemexpress LLC P18IN003 | 71727-40-7 | 99.5% | 296.32 g/mol | C17H16N2O3 | 5MG

Supplier: Medchemexpress LLC HY1235355MG
P18IN003 is a small-molecule inhibitor that selectively targets p18 (INK4C), a cyclin-dependent kinase regulator. It is intended for research use in studies of cell cycle regulation and hematopoietic stem cell expansion, and is characterized with defined physicochemical properties and storage recommendations.
- Selective inhibitor of p18 (INK4C) for research and cell cycle studies.
- Demonstrated potential to promote hematopoietic stem cell expansion.
- CAS 71727-40-7 for unambiguous substance identification.
- Chemical formula C17H16N2O3, molecular weight 296.32 g/mol.
- High purity (99.5%) and white to off-white solid appearance.
- Recommended storage: 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).
- Available in multiple pack sizes including 5 MG, 10 MG, 25 MG, 50 MG, and 100 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.