Learn More
Santa Cruz Biotechnology Anti-p21 Waf1/Cip1/CDKN1A Antibody (F-5), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC6246
Encompass
Anti-p21 Waf1/Cip1/CDKN1A Antibody (F-5) is recommended for detection of p21 Waf1/Cip1 of mouse, rat and human origin by WB, IP, IF, IHC(P) and FCM.
Catalog No. NC1546679