Learn More
Selleck Chemical LLC P22077 10mg 1247819-59-5
SDP
Supplier:  Selleck Chemical LLC S713310MG
Encompass_Preferred
P22077 is an inhibitor of ubiquitin-specific protease USP7 with EC50 of 8.6 ?M, also inhibits the closely related USP47. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4439