Learn More
Aviva Systems Biology Corp P2ry1 Antibody - middle region (ARP51290_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51290_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against P2ry1. It was validated on Western Blot. . Alternate names: P2y, P2y1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-574