Learn More
Medchemexpress LLC P2X3 protein, human (HEK293) | >90.0% | 50UG

Supplier: Medchemexpress LLC HYP70317250UG
Recombinant human P2X3 protein expressed in HEK293 cells and supplied tag-free for research use. Intended for receptor biology studies, biochemical characterization, antibody validation, and assay development.
- Purity greater than 90% as determined by reducing SDS-PAGE.
- Molecular weight approximately 40.4 kDa.
- Tag free formulation.
- Expressed in HEK293 mammalian cells.
- Available in small quantities (10 μg, 50 μg, 100 μg).
- Suitable for biochemical assays and assay development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.