Learn More
Lonza Walkersville P3 Primary Cell 384-well Nucleofector Kit
Supplier:  Lonza Walkersville V5SP3010
Encompass
The P3 Primary Cell 384-well Nucleofector Kit is one of our five kits suited for high-throughput transfection of primary cells when working with the 384-well HT Nucleofector System.Benefits:High transfection efficiencies primary cells - Cells maintain functionality post transfectionSeamless integration into automated liquid handling environmentsConditions are transferable between 4D -ucleofector System, 96-well Shuttle System and 384-well Nucleofector SystemApplications:Transfection of low cell numbers down to 2 x 10⁴ primary cellsHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:90 mL P3 Primary Cell Nucleofector Solution, 20 mL Supplement 1, 50 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0), 10 384-well Nucleocuvette Plate (20 μl)
Catalog No. NC2073405
Only null left
Add to Cart
Request a Quote