Learn More
Medchemexpress LLC HY-124718 5mg Medchemexpress, p32 Inhibitor M36 CAS:802555-85-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1247185MG
Encompass_Preferred
Medchemexpress, HY-124718 5mg p32 Inhibitor M36 CAS:802555-85-7 p32 inhibitor M36 (M36) is a p32 mitochondrial protein inhibitor, which binds directly to p32 and inhibits p32 association with LyP-1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9679