Learn More
Medchemexpress LLC P32 inhibitor M36 | 802555-85-7 | 98.6% | 448.52 | C23H28N8O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12471810MG
Encompass_Preferred
p32 Inhibitor M36 is a small-molecule inhibitor that binds the mitochondrial protein p32 and blocks its association with the tumor-homing peptide LyP-1. It is provided as a purified research reagent for biochemical and cell-based studies of p32 function.
  • Binds directly to p32, inhibiting p32-LyP-1 interaction.
  • Suitable for biochemical and cell-based assays.
  • Available in multiple pack sizes to support varied experimental scales.
  • High purity for reproducible experimental results.
  • Molecular weight 448.52; chemical formula C23H28N8O2.
Catalog No. 50-002-36161