Learn More
Cayman Chemical P3298hemIfumarate 5mg
SDP
Supplier:  Cayman Chemical 168875 MG
Encompass_Preferred
A competitive transition-state substrate analog inhibitor of DPP IV (Ki = 126 nM); used to improve glucose tolerance, insulin sensitivity, and β-cell glucose responsiveness in diabetic rat models at 20 mg/kg/day
Catalog No. 50-296-6757