Learn More
CPC Scientific H-Ala-Asp-Ala-Gln-His-Ala-Thr-Pro-Pro-Lys-Lys-Lys-Arg-Lys-Val-Glu-Asp-Pro-Lys-Asp-Phe-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP020A
Encompass
Specific substrate for p34 cdc2 protein kinase (Km= 0.07 mM), Eliminate multiple phosphorylation sites, leaving only one [specific substrate for p34cdc2 protein kinase (Km= 0.07 mM)] threonine residue available for phosphorylation, thereby reducing the probability of phosphorylation by other protein kinases. ONE-LETTER SEQUENCE: ADAQHATPPKKKRKVEDPKDFMOLECULAR FORMULA: C106H172N32O32MOLECULAR WEIGHT:2406.73STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [135546-44-0], SYNONYMS: CSH 103, Protein Kinase P34 (cd2) SubstrateRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: D.R. Marshak et al., J. Cell. Biochem., 45, 391 (1991); D. McVey et al., Nature, 341, 503 ( 1989); D.R. Marshak et al., J. Cell. Biochem, 45, 391 (1991)
Catalog No. 50-211-0173