Learn More
Medchemexpress LLC p38 alpha/MAPK14 Antibody (YA696) | 50 UL

Supplier: Medchemexpress LLC HYP8077650UL
p38 alpha/MAPK14 Antibody (YA696) is a mouse-derived, non-conjugated IgG1 monoclonal antibody that targets p38 alpha/MAPK14. This antibody is affinity purified and designed for research use, with applications in Western Blot analysis.
- Host: Mouse
- Clonality: Monoclonal
- Molecular weight: Predicted 41 kDa; observed 41 kDa
- Species reactivity: Human, mouse, rat, monkey
- Immunogen: Synthetic peptide corresponding to human P38 MAPK
- Application: WB (Western Blot) 1:500-1:1000
- Sensitivity: Endogenous
- Purity: Affinity purified
- Conjugation: Non-conjugated
- Isotype: IgG1
- Appearance: Liquid
- Formulation: Supplied in 1*PBS (pH 7.3), 50% glycerol and 0.5% BSA; preservative: 0.02% sodium azide
- Storage conditions: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.